Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153416.2 | NP_700465.2 | 5 | 556 | ||
GenBank mRNA | AK034591.1 | - | 5 | 451 | ||
AK079849.1 | - | 5 | 493 | |||
AK083401.1 | - | 5 | 469 | |||
AK083537.1 | - | 5 | 477 | |||
AK087134.1 | - | 5 | 532 | |||
AK088247.1 | - | 5 | 473 | |||
AK134406.1 | - | 5 | 447 | |||
AK165243.1 | - | 5 | 471 | |||
AK166118.1 | - | 5 | 471 | |||
AK169523.1 | - | 5 | 508 | |||
AK189195.1 | - | 4 | 247 | |||
BC018191.1 | - | 5 | 457 | 3994248 | ||
BC025501.1 | - | 5 | 503 | 5321013 | ||
BC026604.1 | - | 4 | 725 |