Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144852.3 | NP_659101.2 | 3 | 1378 | ||
GenBank mRNA | AK030586.1 | - | 3 | 1382 | ||
AK043772.1 | - | 3 | 1379 | |||
AK162398.1 | - | 3 | 1422 |