Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145479.4 | NP_663454.3 | 4 | 649 | ||
GenBank mRNA | AK028197.1 | - | 4 | 1148 | ||
AK030669.1 | - | 4 | 594 | |||
AK086748.1 | - | 4 | 596 | |||
AK143514.1 | - | 4 | 594 | |||
BC005800.1 | - | 4 | 627 | 2812589 |