Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lemd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146075.2 | NP_666187.2 | 7, 8 | 1302 | ||
GenBank mRNA | AK082131.1 | - | 7, 8 | 1109 | ||
AK143224.1 | - | 7, 8 | 1287 | |||
AY640307.1 | - | 7, 8 | 1282 | |||
BC026588.1 | - | 7, 8 | 612 | 4972900 | ||
BC050010.1 | - | 5, 6 | 1540 |