Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013880.3 | NP_038908.2 | 2 | 2194 | ||
GenBank mRNA | AB033615.1 | - | 2 | 2322 | ||
AK122439.1 | - | 1416 | ||||
AK134985.1 | - | 1 | 525 | |||
AK137821.1 | - | 1 | 1104 | |||
AK154288.1 | - | 2 | 2150 | |||
AK165548.1 | - | 2 | 2196 | |||
BC027746.1 | - | 2 | 2256 | 5065782 |