Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ammecr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242430.1 | NP_001229359.1 | 7 | 1510 | ||
NM_153515.4 | NP_705735.1 | 7 | 1309 | |||
GenBank mRNA | AK133018.1 | - | 7 | 1511 | ||
AK151060.1 | - | 7 | 1317 | |||
AK151257.1 | - | 7 | 1217 | |||
BC012310.1 | - | 6 | 914 | |||
BC034661.1 | - | 7 | 1050 | 5137474 | ||
BC056948.1 | - | 7 | 1253 | 6827318 |