Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110015.1 | NP_001103485.1 | 14 | 1687 | ||
NM_001110016.1 | NP_001103486.1 | 14 | 1687 | |||
NM_144863.4 | NP_659112.2 | 14 | 1687 | |||
GenBank mRNA | AK041763.1 | - | 14 | 1607 | ||
AK045396.1 | - | 14 | 1660 | |||
AK077545.1 | - | 14 | 1616 | |||
AK155965.1 | - | 14 | 1585 | |||
AK156822.1 | - | 1686 | ||||
AK171683.1 | - | 14 | 1625 | |||
AK172054.1 | - | 14 | 1612 | |||
AK172377.1 | - | 14 | 1617 | |||
AK186642.1 | - | 3 | 283 | |||
AK213772.1 | - | 1 | 64 | |||
BC022132.1 | - | 14 | 1659 | |||
BC025047.1 | - | 1 | 45 | |||
BC025809.1 | - | 14 | 1601 | |||
BC094453.1 | - | 14 | 1638 | 5068941 |