Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153388.4 | NP_700437.2 | 2 | 1914 | ||
GenBank mRNA | AK081560.1 | - | 1 | 2197 | ||
AK163603.1 | - | 2 | 1772 | |||
AK214339.1 | - | 1 | 318 | |||
BC023036.1 | - | 1 | 1527 | 5358628 | ||
BC023156.1 | - | 2 | 1570 | 4951386 | ||
DQ078786.1 | - | 1 | 1199 |