Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001984.2 | NP_001001984.2 | 11, 12 | 1913 | ||
GenBank mRNA | AK155866.1 | - | 11, 12 | 1918 | ||
BC037117.1 | - | 4, 5 | 462 | |||
BC076576.1 | - | 11, 12 | 1479 | 6840591 |