Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005507.2 | NP_001005507.1 | 14 | 1821 | ||
GenBank mRNA | BC082789.1 | - | 14 | 1808 | 30105222 |