Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144878.1 | NP_659127.1 | 6 | 725 | ||
GenBank mRNA | AF461145.1 | - | 6 | 725 | ||
AK146562.1 | - | 7 | 928 |