Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081386.1 | NP_001074855.1 | 7 | 1232 | ||
GenBank mRNA | AK148396.1 | - | 7 | 1220 | ||
BC157927.1 | - | 7 | 1223 | 9088100 |