Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177647.4 | NP_808315.1 | 2 | 191 | ||
GenBank mRNA | AK034009.1 | - | 2 | 193 | ||
BC110560.2 | - | 2 | 179 | 40049634 |