Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048176.1 | NP_001041641.1 | 11 | 1400 | ||
GenBank mRNA | BC046474.1 | - | 11 | 1506 | ||
CJ235603.1 | - | 5 | 331 | |||
HQ426680.1 | - | 11 | 1400 | |||
HQ426681.1 | - | 9 | 1193 | |||
HQ426682.1 | - | 8 | 1061 | |||
HQ426683.1 | - | 7 | 818 | |||
HQ426684.1 | - | 10 | 1268 | |||
HQ426685.1 | - | 10 | 1340 | |||
HQ426686.1 | - | 10 | 1257 | |||
HQ426687.1 | - | 11 | 1531 | |||
HQ426688.1 | - | 11 | 1547 | |||
HQ426689.1 | - | 10 | 1415 | |||
HQ426690.1 | - | 11 | 1617 | |||
HQ426691.1 | - | 1199 | ||||
HQ426692.1 | - | 1330 | ||||
HQ426693.1 | - | 1240 | ||||
HQ426694.1 | - | 1153 | ||||
HQ426695.1 | - | 1372 | ||||
HQ426696.1 | - | 1312 | ||||
HQ426697.1 | - | 1457 | ||||
HQ426698.1 | - | 1165 | ||||
HQ426699.1 | - | 1229 | ||||
HQ426700.1 | - | 1503 | ||||
HQ426701.1 | - | 1519 | ||||
HQ426702.1 | - | 10 | 982 | |||
HQ426703.1 | - | 8 | 775 | |||
HQ426704.1 | - | 10 | 982 | |||
HQ426705.1 | - | 9 | 839 | |||
HQ426706.1 | - | 9 | 926 | |||
HQ426707.1 | - | 9 | 1073 | |||
HQ426708.1 | - | 5 | 436 | |||
HQ426709.1 | - | 7 | 674 | |||
HQ426710.1 | - | 6 | 614 | |||
HQ426711.1 | - | 7 | 821 |