Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145832.3 | NP_665831.1 | 2 | 509 | ||
NM_211358.2 | NP_997597.1 | 1 | 997 | |||
GenBank mRNA | AJ440720.1 | - | 1 | 394 | ||
AK030977.1 | - | 2 | 510 | |||
AK033295.1 | - | 1 | 86 | |||
AK040986.1 | - | 1 | 744 | |||
AK045972.1 | - | 2 | 486 | |||
AK050311.1 | - | 2 | 474 | |||
AK081068.1 | - | 1 | 495 | |||
AK088227.1 | - | 2 | 496 | |||
AK138595.1 | - | 1 | 940 | |||
AK144762.1 | - | 1 | 919 | |||
AK151154.1 | - | 1 | 993 | |||
BC031814.1 | - | 1 | 591 | 4196818 | ||
BC116736.1 | - | 1 | 422 | 40126055 | ||
BC119128.1 | - | 1 | 422 | 8733877 |