Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178695.5 | NP_848810.3 | 3 | 548 | ||
GenBank mRNA | AK035530.1 | - | 3 | 373 | ||
AK037087.1 | - | 3 | 468 | |||
AK038703.1 | - | 3 | 548 | |||
AK166405.1 | - | 2 | 509 | |||
BC099390.1 | - | 3 | 465 | 30715383 | ||
BC115470.2 | - | 3 | 294 | 40100436 | ||
BC151011.1 | - | 3 | 478 | 9087924 | ||
BC151019.1 | - | 3 | 478 | 9087932 |