Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045523.1 | NP_001038988.1 | 2 | 540 | ||
GenBank mRNA | AK129243.1 | - | 2 | 698 | ||
AK152597.1 | - | 2 | 478 | |||
BC100358.1 | - | 2 | 702 | 30848279 | ||
BC141046.1 | - | 1 | 403 | 9055658 | ||
BC145233.1 | - | 1 | 403 | 9053785 |