Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172269.3 | NP_758473.3 | 5 | 2723 | ||
GenBank mRNA | AK029109.1 | - | 5 | 2700 | ||
AK033333.1 | - | 5 | 2708 | |||
AK036915.1 | - | 5 | 2708 | |||
AK036947.1 | - | 2 | 886 | |||
AK039848.1 | - | 5 | 2707 | |||
AK042471.1 | - | 5 | 2711 | |||
AK148077.1 | - | 5 | 2715 | |||
AK151113.1 | - | 2 | 1345 | |||
AK152700.1 | - | 2 | 1345 | |||
AK160689.1 | - | 5 | 2701 | |||
AK173179.1 | - | 2 | 1869 | |||
AK205924.1 | - | 1 | 120 | |||
BC026870.1 | - | 2 | 1649 | |||
BC036129.1 | - | 5 | 2651 | 4972693 | ||
BC039176.1 | - | 5 | 2630 | 4221232 |
Gene Symbol: Atp6v0c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK189660.1 | - | 119 |