Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ythdf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173761.3 | NP_776122.1 | 4 | 1375 | ||
GenBank mRNA | AK033241.1 | - | 5 | 1466 | ||
AK046768.1 | - | 4 | 1389 | |||
AK075846.1 | - | 4 | 1354 | |||
AK172582.1 | - | 4 | 1349 | |||
AK192799.1 | - | 1 | 171 | |||
AK205485.1 | - | 1 | 66 | |||
BC061479.1 | - | 4 | 1346 | 6334535 | ||
BC065050.1 | - | 4 | 1375 | 6418891 |