Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183162.2 | NP_898985.2 | 9 | 6274 | ||
GenBank mRNA | AB201714.1 | - | 8 | 6178 | ||
AK173259.1 | - | 1 | 3194 |