Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172676.2 | NP_766264.2 | 3, 4 | 581 | ||
GenBank mRNA | AK053048.1 | - | 3, 4 | 535 | ||
AK053884.1 | - | 1, 2 | 162 | |||
AK164204.1 | - | 3, 4 | 620 | |||
AK171151.1 | - | 3, 4 | 587 | |||
BC019475.1 | - | 3, 4 | 591 | |||
BC052921.1 | - | 3, 4 | 581 | 6311715 |