Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172678.3 | NP_766266.3 | 18 | 1938 | ||
GenBank mRNA | AK012773.1 | - | 5 | 463 | ||
AK028571.1 | - | 17 | 2652 | |||
AK031820.1 | - | 18 | 1870 | |||
AK075984.1 | - | 18 | 1857 | |||
AK088409.1 | - | 5 | 373 | |||
AK145423.1 | - | 18 | 1846 | |||
AK184631.1 | - | 3 | 165 | |||
AK209821.1 | - | 3 | 164 | |||
BC004737.1 | - | 11 | 1827 | |||
BC031137.1 | - | 18 | 1874 | 4971848 | ||
BC032213.1 | - | 18 | 1879 | 5323723 | ||
BC033277.1 | - | 18 | 1859 | 4241041 |