Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033301.4 | NP_001028473.2 | 4 | 792 | ||
NM_001205355.1 | NP_001192284.1 | 4 | 863 | |||
GenBank mRNA | AK031946.1 | - | 3 | 268 | ||
AK145217.1 | - | 4 | 783 | |||
AK173244.1 | - | 4 | 863 | |||
BC042779.1 | - | 5 | 478 | |||
BC137632.1 | - | 4 | 832 | 8860651 |