Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177663.4 | NP_808331.1 | 4 | 1515 | ||
GenBank mRNA | AK053778.1 | - | 4 | 1516 | ||
AK076747.1 | - | 4 | 1238 | |||
AK155979.1 | - | 4 | 1519 | |||
AK178428.1 | - | 2 | 149 | |||
BC098326.1 | - | 4 | 1502 | 5720443 |