Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146131.2 | NP_666243.1 | 10 | 1691 | ||
GenBank mRNA | AK028754.1 | - | 10 | 1652 | ||
AK028858.1 | - | 10 | 1689 | |||
AK159422.1 | - | 10 | 1678 | |||
AK159448.1 | - | 10 | 1678 | |||
AK159633.1 | - | 10 | 1668 | |||
AK160103.1 | - | 10 | 1672 | |||
AK170318.1 | - | 10 | 1647 | |||
BC026838.1 | - | 9 | 1626 | 4460993 | ||
BC027771.1 | - | 10 | 1661 | 5344900 | ||
BC034200.1 | - | 10 | 1632 | 4986595 |