Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gatad2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139304.1 | NP_647465.1 | 10 | 1731 | ||
GenBank mRNA | AF411837.1 | - | 10 | 1731 | ||
AK041594.1 | - | 10 | 2952 | |||
AK049450.1 | - | 2 | 181 | |||
AK173113.1 | - | 7 | 1049 | |||
BC060294.1 | - | 5 | 525 | |||
BC130262.1 | - | 10 | 1685 | 40142858 |