Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145540.3 | NP_663515.2 | 10 | 1728 | ||
NM_178876.3 | NP_849207.2 | 10 | 1728 | |||
GenBank mRNA | AK033296.1 | - | 10 | 1395 | ||
AK146699.1 | - | 10 | 1622 | |||
AK147865.1 | - | 10 | 1707 | |||
BC055344.1 | - | 10 | 1263 | 6827687 |