Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030016.2 | NP_084292.1 | 2, 3 | 1792 | ||
GenBank mRNA | AK039588.1 | - | 2, 3 | 1793 | ||
AK041636.1 | - | 1, 2 | 258 | |||
AK163561.1 | - | 1, 2 | 207 | |||
BC127174.1 | - | 1, 2 | 150 |