Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rap1gds1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040690.2 | NP_001035780.1 | 4 | 453 | ||
NM_001286759.1 | NP_001273688.1 | 4 | 453 | |||
NM_145544.3 | NP_663519.2 | 4 | 453 | |||
GenBank mRNA | AK052125.1 | - | 4 | 401 | ||
AK154852.1 | - | 4 | 439 | |||
AK155793.1 | - | 4 | 439 | |||
AK158465.1 | - | 4 | 446 | |||
AK160989.1 | - | 4 | 389 | |||
AK164159.1 | - | 4 | 365 | |||
AK166315.1 | - | 4 | 386 | |||
AK172052.1 | - | 4 | 455 | |||
BC011279.1 | - | 4 | 322 | 4008751 |