Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290645.1 | NP_001277574.1 | 6 | 998 | ||
NM_146148.2 | NP_666260.1 | 7 | 1095 | |||
GenBank mRNA | AB077295.1 | - | 7 | 1109 | ||
AB077296.1 | - | 7 | 1058 | |||
AB077297.1 | - | 6 | 964 | |||
AB077298.1 | - | 7 | 1058 | |||
AK090245.1 | - | 7 | 1095 | |||
BC027748.1 | - | 7 | 1091 | 5052412 |