Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175472.3 | NP_780681.2 | 6 | 1624 | ||
GenBank mRNA | AK031299.1 | - | 4 | 1017 | ||
AK041821.1 | - | 6 | 1630 | |||
AK049279.1 | - | 4 | 613 | |||
AK220327.1 | - | 4 | 872 | |||
BC145507.1 | - | 5 | 1275 | |||
BC150791.1 | - | 5 | 1275 | 9087702 |