Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK028000.1 | - | 1 | 24 | ||
AK052762.1 | - | 4 | 1327 | |||
AK085717.1 | - | 5 | 1489 | |||
AK154613.1 | - | 4 | 1473 | |||
AK163171.1 | - | 4 | 1420 | |||
AK170641.1 | - | 4 | 1459 | |||
AK180272.1 | - | 1 | 99 | |||
AK181367.1 | - | 1 | 277 | |||
AK202629.1 | - | 1 | 99 | |||
AK210294.1 | - | 1 | 99 | |||
BC005765.1 | - | 3 | 1195 | |||
BC007169.1 | - | 1 | 352 | |||
BC022976.1 | - | 4 | 1405 | 5362925 | ||
BC024940.1 | - | 4 | 1402 | |||
BC071258.1 | - | 4 | 1338 |