Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmpste24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172700.2 | NP_766288.1 | 4 | 675 | ||
GenBank mRNA | AJ487544.1 | - | 4 | 641 | ||
AK083566.1 | - | 4 | 675 | |||
AK170946.1 | - | 4 | 633 | |||
AY029194.1 | - | 4 | 431 | |||
BC023740.1 | - | 4 | 617 | |||
BC138578.1 | - | 4 | 543 | 8861598 |