Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161851.2 | NP_001155323.1 | 6 | 1308 | ||
NM_001161852.2 | NP_001155324.1 | 5 | 1460 | |||
NM_173401.5 | NP_775577.2 | 6 | 1585 | |||
GenBank mRNA | AK077452.1 | - | 6 | 1371 | ||
AK170958.1 | - | 7 | 1352 | |||
AK182933.1 | - | 1 | 375 | |||
AK184124.1 | - | 1 | 346 | |||
AK188052.1 | - | 1 | 351 | |||
AK188531.1 | - | 1 | 356 | |||
AK218373.1 | - | 1 | 119 | |||
AK218416.1 | - | 1 | 324 | |||
BY658029.1 | - | 1 | 146 |