Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172704.3 | NP_766292.2 | 3 | 265 | ||
GenBank mRNA | AK053156.1 | - | 3 | 247 | ||
AK089983.1 | - | 3 | 257 | |||
AK156793.1 | - | 3 | 247 | |||
AK171781.1 | - | 3 | 246 | |||
AK172542.1 | - | 3 | 246 | |||
BC075619.1 | - | 3 | 241 | |||
BC079852.1 | - | 3 | 235 | |||
BC085257.1 | - | 3 | 239 | 5414963 | ||
BY784439.1 | - | 3 | 276 |