Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cc2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172274.2 | NP_758478.1 | 8 | 947 | ||
GenBank mRNA | AK077658.1 | - | 5 | 422 | ||
AK145128.1 | - | 5 | 485 | |||
BC032290.1 | - | 8 | 891 | 5369681 |