Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144919.2 | NP_659168.1 | 9 | 698 | ||
GenBank mRNA | AK045994.1 | - | 9 | 710 | ||
BC016208.1 | - | 9 | 698 | 4913322 |