Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205363.1 | NP_001192292.1 | 6 | 655 | ||
NM_001205364.1 | NP_001192293.1 | 5 | 574 | |||
NM_172732.3 | NP_766320.2 | 5 | 577 | |||
GenBank mRNA | AK036399.1 | - | 6 | 656 | ||
AK041288.1 | - | 5 | 577 | |||
BC087955.1 | - | 6 | 654 | 30119997 | ||
EU339277.1 | - | 5 | 484 | |||
EU339278.1 | - | 6 | 562 | |||
EU339279.1 | - | 5 | 481 |