Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301407.1 | NP_001288336.1 | 6 | 721 | ||
NM_146173.3 | NP_666285.1 | 6 | 724 | |||
GenBank mRNA | AF276890.1 | - | 6 | 723 | ||
AK004954.1 | - | 6 | 697 | |||
AK172176.1 | - | 6 | 702 | |||
BC024685.1 | - | 6 | 682 | 3981492 | ||
BE861472.1 | - | 4 | 333 | |||
BU054333.1 | - | 6 | 475 |