Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171003.1 | NP_001164474.1 | 47, 48 | 5581 | ||
GenBank mRNA | AK031332.1 | - | 31, 32 | 3584 | ||
AK033548.1 | - | 7, 8 | 720 | |||
BC025509.1 | - | 13, 14 | 1281 | |||
EU937529.1 | - | 47, 48 | 5581 |