Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172736.3 | NP_766324.1 | 14 | 2131 | ||
GenBank mRNA | AK034310.1 | - | 14 | 2136 | ||
AK170629.1 | - | 14 | 2139 | |||
BC034448.1 | - | 6 | 855 | |||
BC041775.1 | - | 13 | 2636 | |||
BC042658.1 | - | 14 | 2119 | 5365041 | ||
BC066768.1 | - | 14 | 2122 | 6407146 |