Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199060.1 | NP_001185989.1 | 9 | 1270 | ||
NM_001199061.1 | NP_001185990.1 | 9 | 1002 | |||
NM_021312.5 | NP_067287.1 | 8 | 1092 | |||
GenBank mRNA | AB041608.1 | - | 9 | 1271 | ||
AF239765.1 | - | 8 | 748 | |||
AK007500.1 | - | 8 | 869 | |||
AK016611.1 | - | 9 | 946 | |||
AK031436.1 | - | 8 | 855 | |||
AK149085.1 | - | 8 | 774 | |||
AK161299.1 | - | 9 | 1015 | |||
AY059431.1 | - | 8 | 783 | |||
AY059432.1 | - | 1 | 88 | |||
BC004748.1 | - | 8 | 926 | 3583955 | ||
BC052386.1 | - | 8 | 1063 | 6418498 | ||
BC096651.1 | - | 8 | 837 | 1514703 |