Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198613.2 | NP_941015.2 | 3 | 309 | ||
GenBank mRNA | AK146584.1 | - | 3 | 310 | ||
AK186335.1 | - | 1 | 61 | |||
AK206960.1 | - | 1 | 61 | |||
BC023066.1 | - | 1 | 561 | |||
BC052499.1 | - | 4 | 270 | 3370978 |