Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nccrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081115.1 | NP_001074584.1 | 2, 3 | 568 | ||
GenBank mRNA | AK028022.1 | - | 2, 3 | 218 |