Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2af1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170760.3 | NP_739566.1 | 2 | 127 | ||
GenBank mRNA | AF419339.1 | - | 2 | 111 | ||
BC051477.1 | - | 2 | 135 | |||
BC060972.1 | - | 2 | 127 | 6593404 |