Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002687.3 | NP_002678.2 | 5 | 472 | ||
GenBank mRNA | AK223612.1 | - | 5 | 407 | ||
AK292579.1 | - | 5 | 456 | |||
AK303136.1 | - | 1 | 63 | |||
BC062602.1 | - | 5 | 383 | 5491466 | ||
U77718.1 | - | 5 | 402 | |||
Y09703.1 | - | 5 | 382 |