Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173767.3 | NP_776128.2 | 10 | 1509 | ||
GenBank mRNA | AK050143.1 | - | 7 | 1004 | ||
AK178410.1 | - | 2 | 183 | |||
AK196619.1 | - | 2 | 183 | |||
BC026405.1 | - | 10 | 1396 | 4211657 | ||
BC089586.1 | - | 7 | 1013 | |||
BC139196.1 | - | 7 | 897 | 8862218 | ||
BC139197.1 | - | 7 | 897 | 8862219 | ||
DQ205645.1 | - | 9 | 1222 |