Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031814.1 | NP_001026984.1 | 7 | 1259 | ||
GenBank mRNA | AK041264.1 | - | 7 | 1038 | ||
AK047829.1 | - | 7 | 1240 | |||
AK049391.1 | - | 7 | 1259 | |||
AK157798.1 | - | 7 | 1246 |