Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tufm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163713.1 | NP_001157185.1 | 9 | 1396 | ||
NM_172745.3 | NP_766333.1 | 10 | 1308 | |||
GenBank mRNA | AK075857.1 | - | 10 | 1232 | ||
AK084724.1 | - | 10 | 1289 | |||
AK152858.1 | - | 10 | 1303 | |||
AK153135.1 | - | 10 | 1303 | |||
BC060959.1 | - | 9 | 1373 | 30379757 | ||
BC100596.1 | - | 10 | 1301 | 6762419 |