Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hirip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172746.3 | NP_766334.1 | 4 | 1498 | ||
GenBank mRNA | AK045122.1 | - | 4 | 1033 | ||
AK135526.1 | - | 4 | 1043 | |||
AK161657.1 | - | 4 | 1033 | |||
AK162096.1 | - | 1 | 60 | |||
BC055687.1 | - | 4 | 1498 | 6414655 |